Assay Details
| Gene Symbol: chrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113043.1 | NP_001106514.1 | 1 | 1256 | 94 | |
| GenBank mRNA | BC122953.1 | - | 1 | 1327 | 94 | |
| BC154720.1 | - | 1 | 1256 | 94 | ||
More Information
Set Membership: |
|

chrm4
Genomic Map