Assay Details
| Gene Symbol: hla-a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113065.1 | NP_001106536.1 | 1849 | 99 | ||
| GenBank mRNA | AY204556.1 | - | 3 | 960 | 99 | |
| AY204559.1 | - | 1851 | 99 | |||
| BC154904.1 | - | 1849 | 99 | |||
More Information
Set Membership: |
|

hla-a
Genomic Map