Assay Details
| Gene Symbol: hla-dqa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113067.1 | NP_001106538.1 | 748 | 113 | ||
| GenBank mRNA | BC154907.1 | - | 748 | 113 | ||
| BC168592.1 | - | 1 | 449 | 113 | ||
More Information
Set Membership: |
|

hla-dqa1
Genomic Map