Assay Details
| Gene Symbol: fam43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114027.1 | NP_001107499.1 | 1 | 746 | 85 | |
| GenBank mRNA | BC157653.1 | - | 1 | 746 | 85 | |
| BC171115.1 | - | 1 | 668 | 85 | ||
| BC171119.1 | - | 1 | 668 | 85 | ||
More Information
Set Membership: |
|

fam43a
Genomic Map