Assay Details
| Gene Symbol: btn2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130216.1 | NP_001123688.1 | 253 | 88 | ||
| GenBank mRNA | BC166273.1 | - | 253 | 88 | ||
| Gene Symbol: LOC100495374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018089754.1 | XP_017945243.1 | 2 | 246 | 88 | |
More Information
Set Membership: |
|

btn2a1
Genomic Map