Assay Details
| Gene Symbol: inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134805.1 | NP_001128277.1 | 3 | 750 | 85 | |
| GenBank mRNA | BC091096.1 | - | 3 | 728 | 85 | |
| BC123980.1 | - | 3 | 728 | 85 | ||
| BC154880.1 | - | 3 | 748 | 85 | ||
| BC169145.1 | - | 3 | 750 | 85 | ||
More Information
Set Membership: |
|

inmt
Genomic Map