Assay Details
| Gene Symbol: vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203574.1 | NP_988905.1 | 254 | 96 | ||
| GenBank mRNA | BC061623.1 | - | 254 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vapb| Gene Symbol: vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203574.1 | NP_988905.1 | 254 | 96 | ||
| GenBank mRNA | BC061623.1 | - | 254 | 96 | ||
Set Membership: |
|