Assay Details
| Gene Symbol: sox17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204094.1 | NP_989425.1 | 2 | 1153 | 112 | |
| GenBank mRNA | AY157635.1 | - | 2 | 1153 | 112 | |
| BC074527.1 | - | 2 | 112 | |||
| CR848309.2 | - | 2 | 112 | |||
More Information
Set Membership: |
|

sox17a
Genomic Map