Assay Details
| Gene Symbol: LOC100216713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143118.1 | NP_001136590.1 | 283 | 78 | ||
| GenBank mRNA | BT037225.1 | - | 283 | 78 | ||
| EU954392.1 | - | 328 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100216713| Gene Symbol: LOC100216713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143118.1 | NP_001136590.1 | 283 | 78 | ||
| GenBank mRNA | BT037225.1 | - | 283 | 78 | ||
| EU954392.1 | - | 328 | 78 | |||