Assay Details
| Gene Symbol: LOC100216713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143118.1 | NP_001136590.1 | 389 | 70 | ||
| GenBank mRNA | BT037225.1 | - | 389 | 70 | ||
| EU954392.1 | - | 434 | 70 | |||
More Information
Set Membership: |
|

LOC100216713
Genomic Map