Assay Details
| Gene Symbol: LOC100274124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148502.1 | NP_001141974.1 | 1005 | 71 | ||
| GenBank mRNA | BT042400.1 | - | 1005 | 71 | ||
More Information
Set Membership: |
|

LOC100274124
Genomic Map