Assay Details
| Gene Symbol: LOC100274194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148567.1 | NP_001142039.1 | 1409 | 58 | ||
| GenBank mRNA | BT042514.1 | - | 1409 | 58 | ||
More Information
Set Membership: |
|

LOC100274194
Genomic Map