Assay Details
| Gene Symbol: LOC100281095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154014.1 | NP_001147486.1 | 372 | 112 | ||
| GenBank mRNA | EU955563.1 | - | 372 | 112 | ||
More Information
Set Membership: |
|

LOC100281095
Genomic Map