Assay Details
| Gene Symbol: LOC100282181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155093.1 | NP_001148565.1 | 652 | 102 | ||
| GenBank mRNA | EU959940.1 | - | 652 | 102 | ||
More Information
Set Membership: |
|

LOC100282181
Genomic Map