Assay Details
| Gene Symbol: LOC100283544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156444.1 | NP_001149916.1 | 1105 | 75 | ||
| GenBank mRNA | EU965067.1 | - | 1105 | 75 | ||
More Information
Set Membership: |
|

LOC100283544
Genomic Map