Assay Details
| Gene Symbol: LOC100284061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156959.1 | NP_001150431.1 | 1283 | 98 | ||
| GenBank mRNA | EU966955.1 | - | 1283 | 98 | ||
More Information
Set Membership: |
|

LOC100284061
Genomic Map