Assay Details
| Gene Symbol: BE997321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157913.1 | NP_001151385.1 | 786 | 91 | ||
| GenBank mRNA | EU970525.1 | - | 786 | 91 | ||
| Gene Symbol: LOC100381822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174617.1 | NP_001168088.1 | 321 | 91 | ||
| GenBank mRNA | BT062356.1 | - | 321 | 91 | ||
More Information
Set Membership: |
|

BE997321
Genomic Map