Assay Details
| Gene Symbol: LOC100285121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158016.1 | NP_001151488.1 | 1058 | 74 | ||
| GenBank mRNA | EU970930.1 | - | 1058 | 74 | ||
More Information
Set Membership: |
|

LOC100285121
Genomic Map