Assay Details
| Gene Symbol: LOC100285561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158452.1 | NP_001151924.1 | 916 | 83 | ||
| GenBank mRNA | EU972907.1 | - | 916 | 83 | ||
More Information
Set Membership: |
|

LOC100285561
Genomic Map