Assay Details
| Gene Symbol: LOC100286031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158919.1 | NP_001152391.1 | 1348 | 106 | ||
| GenBank mRNA | EU975260.1 | - | 1348 | 106 | ||
More Information
Set Membership: |
|

LOC100286031
Genomic Map