Assay Details
| Gene Symbol: ids1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111434.1 | NP_001104904.1 | 1313 | 96 | ||
| GenBank mRNA | AF048900.1 | - | 1313 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ids1| Gene Symbol: ids1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111434.1 | NP_001104904.1 | 1313 | 96 | ||
| GenBank mRNA | AF048900.1 | - | 1313 | 96 | ||
Set Membership: |
|