Assay Details
| Gene Symbol: rop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111459.1 | NP_001104929.1 | 622 | 95 | ||
| GenBank mRNA | AF126052.1 | - | 622 | 95 | ||
| AY106460.1 | - | 837 | 95 | |||
| BT087642.1 | - | 574 | 95 | |||
More Information
Set Membership: |
|

rop1
Genomic Map