Assay Details
| Gene Symbol: rop3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111460.1 | NP_001104930.1 | 425 | 81 | ||
| GenBank mRNA | AF126054.1 | - | 425 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rop3| Gene Symbol: rop3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111460.1 | NP_001104930.1 | 425 | 81 | ||
| GenBank mRNA | AF126054.1 | - | 425 | 81 | ||
Set Membership: |
|