Assay Details
| Gene Symbol: SRP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111793.1 | NP_001105263.1 | 755 | 91 | ||
| GenBank mRNA | AY620403.1 | - | 755 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRP1A| Gene Symbol: SRP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111793.1 | NP_001105263.1 | 755 | 91 | ||
| GenBank mRNA | AY620403.1 | - | 755 | 91 | ||
Set Membership: |
|