Assay Details
| Gene Symbol: SRP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111793.1 | NP_001105263.1 | 1096 | 77 | ||
| GenBank mRNA | AY620402.1 | - | 581 | 77 | ||
| AY620403.1 | - | 1096 | 77 | |||
| AY620404.1 | - | 931 | 77 | |||
| AY620405.1 | - | 832 | 77 | |||
More Information
Set Membership: |
|

SRP1A
Genomic Map