Assay Details
| Gene Symbol: mtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112029.1 | NP_001105499.1 | 147 | 82 | ||
| GenBank mRNA | AY106088.1 | - | 224 | 82 | ||
| EU952603.1 | - | 231 | 82 | |||
| EU955334.1 | - | 233 | 82 | |||
| EU976520.1 | - | 232 | 82 | |||
| U10696.1 | - | 147 | 82 | |||
| Z34469.1 | - | 147 | 82 | |||
More Information
Set Membership: |
|

mtl2
Genomic Map