Assay Details
| Gene Symbol: gla3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112201.1 | NP_001105671.1 | 284 | 81 | ||
| GenBank mRNA | AY344632.1 | - | 284 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gla3| Gene Symbol: gla3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112201.1 | NP_001105671.1 | 284 | 81 | ||
| GenBank mRNA | AY344632.1 | - | 284 | 81 | ||
Set Membership: |
|