Assay Details
| Gene Symbol: LOC100174972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134283.1 | NP_001127755.1 | 1273 | 72 | ||
| GenBank mRNA | EU853674.1 | - | 1273 | 72 | ||
More Information
Set Membership: |
|

LOC100174972
Genomic Map