Assay Details
| Gene Symbol: LOC100191518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136950.1 | NP_001130422.1 | 1208 | 62 | ||
| GenBank mRNA | BT033618.1 | - | 1208 | 62 | ||
| BT066940.1 | - | 1209 | 62 | |||
More Information
Set Membership: |
|

LOC100191518
Genomic Map