Assay Details
| Gene Symbol: spr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137102.1 | NP_001130574.1 | 1054 | 93 | ||
| GenBank mRNA | BT033842.1 | - | 1054 | 93 | ||
| BT042981.1 | - | 1069 | 93 | |||
More Information
Set Membership: |
|

spr1
Genomic Map