Assay Details
| Gene Symbol: LOC100191756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137183.1 | NP_001130655.1 | 1404 | 84 | ||
| GenBank mRNA | BT033956.1 | - | 1404 | 84 | ||
| EU957961.1 | - | 1318 | 84 | |||
More Information
Set Membership: |
|

LOC100191756
Genomic Map