Assay Details
| Gene Symbol: LOC100191828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137252.1 | NP_001130724.1 | 678 | 67 | ||
| GenBank mRNA | BT034055.1 | - | 678 | 67 | ||
| BT034924.1 | - | 737 | 67 | |||
| BT063486.1 | - | 580 | 67 | |||
| EU957181.1 | - | 670 | 67 | |||
More Information
Set Membership: |
|

LOC100191828
Genomic Map