Assay Details
| Gene Symbol: cad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137309.1 | NP_001130781.1 | 322 | 71 | ||
| GenBank mRNA | BT034128.1 | - | 322 | 71 | ||
| BT035050.1 | - | 244 | 71 | |||
| BT040774.1 | - | 319 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT035050.1 transcripts.
Set Membership: |
|

cad1
Genomic Map