Assay Details
| Gene Symbol: cad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137309.1 | NP_001130781.1 | 992 | 67 | ||
| GenBank mRNA | BT034128.1 | - | 992 | 67 | ||
| BT035050.1 | - | 914 | 67 | |||
| BT040774.1 | - | 989 | 67 | |||
More Information
Set Membership: |
|

cad1
Genomic Map