Assay Details
| Gene Symbol: sad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137318.1 | NP_001130790.1 | 400 | 84 | ||
| GenBank mRNA | BT034140.1 | - | 400 | 84 | ||
| BT036418.1 | - | 199 | 84 | |||
More Information
Set Membership: |
|

sad1
Genomic Map