Assay Details
| Gene Symbol: LOC100191906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137330.1 | NP_001130802.1 | 752 | 82 | ||
| GenBank mRNA | BT034107.1 | - | 559 | 82 | ||
| BT034154.1 | - | 696 | 82 | |||
| BT067431.1 | - | 690 | 82 | |||
| EU965164.1 | - | 752 | 82 | |||

LOC100191906
Genomic Map