Assay Details
| Gene Symbol: LOC100192094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137517.1 | NP_001130989.1 | 1265 | 64 | ||
| GenBank mRNA | BT034400.1 | - | 1179 | 64 | ||
| BT063300.1 | - | 1163 | 64 | |||
| EU964300.1 | - | 1265 | 64 | |||
More Information
Set Membership: |
|

LOC100192094
Genomic Map