Assay Details
| Gene Symbol: LOC100272517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146986.1 | NP_001140458.1 | 498 | 76 | ||
| GenBank mRNA | BT038877.1 | - | 498 | 76 | ||
| EU961934.1 | - | 517 | 76 | |||
More Information
Set Membership: |
|

LOC100272517
Genomic Map