Assay Details
Gene Symbol: LOC100273233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147675.1 | NP_001141147.1 | 1035 | 105 | ||
GenBank mRNA | BT040520.1 | - | 959 | 105 | ||
BT067342.1 | - | 1035 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
