Assay Details
| Gene Symbol: LOC100274580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148933.1 | NP_001142405.1 | 1408 | 109 | ||
| GenBank mRNA | BT043409.1 | - | 1408 | 109 | ||
| BT069155.1 | - | 832 | 109 | |||
| EU967749.1 | - | 892 | 109 | |||
More Information
Set Membership: |
|

LOC100274580
Genomic Map