Assay Details
Gene Symbol: LOC100280498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153418.1 | NP_001146890.1 | 1882 | 66 | ||
GenBank mRNA | EU952098.1 | - | 1882 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
