Assay Details
| Gene Symbol: LOC100280703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153622.1 | NP_001147094.1 | 497 | 71 | ||
| GenBank mRNA | EU953301.1 | - | 497 | 71 | ||
More Information
Set Membership: |
|

LOC100280703
Genomic Map