Assay Details
Gene Symbol: LOC100280703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153622.1 | NP_001147094.1 | 879 | 63 | ||
GenBank mRNA | EU953301.1 | - | 879 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
