Assay Details
| Gene Symbol: LOC100280985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153905.1 | NP_001147377.1 | 408 | 66 | ||
| GenBank mRNA | BT084559.1 | - | 499 | 66 | ||
| EU955053.1 | - | 408 | 66 | |||
More Information
Set Membership: |
|

LOC100280985
Genomic Map