Assay Details
| Gene Symbol: LOC100281424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154342.1 | NP_001147814.1 | 1788 | 76 | ||
| GenBank mRNA | EU956665.1 | - | 1788 | 76 | ||
More Information
Set Membership: |
|

LOC100281424
Genomic Map