Assay Details
Gene Symbol: LOC100281424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154342.1 | NP_001147814.1 | 1871 | 80 | ||
GenBank mRNA | EU956665.1 | - | 1871 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
