Assay Details
| Gene Symbol: LOC100281512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154430.1 | NP_001147902.1 | 1466 | 72 | ||
| GenBank mRNA | EU956950.1 | - | 1466 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100281512| Gene Symbol: LOC100281512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154430.1 | NP_001147902.1 | 1466 | 72 | ||
| GenBank mRNA | EU956950.1 | - | 1466 | 72 | ||