Assay Details
| Gene Symbol: LOC100283984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156882.1 | NP_001150354.1 | 1045 | 78 | ||
| GenBank mRNA | EU966657.1 | - | 1045 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100283984| Gene Symbol: LOC100283984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156882.1 | NP_001150354.1 | 1045 | 78 | ||
| GenBank mRNA | EU966657.1 | - | 1045 | 78 | ||