Assay Details
| Gene Symbol: LOC100284031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156929.1 | NP_001150401.1 | 755 | 96 | ||
| GenBank mRNA | EU966832.1 | - | 755 | 96 | ||
More Information
Set Membership: |
|

LOC100284031
Genomic Map