Assay Details
| Gene Symbol: LOC100284150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157047.1 | NP_001150519.1 | 248 | 69 | ||
| GenBank mRNA | EU967269.1 | - | 248 | 69 | ||
More Information
Set Membership: |
|

LOC100284150
Genomic Map